Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
iFluor® 710, Succinimidyl Ester

iFluor® 710, Succinimidyl Ester

1 mg
Catalog # AATB1045
Supplier:   AAT Bioquest
iFluor® 710, Succinimidyl Ester
Catalog # AATB1045
Supplier:  AAT Bioquest
CAS Number:  
iFluor® 710, Succinimidyl Ester
iFluor® 710, Succinimidyl Ester

1 mg

CAS Number:  
Price
2.350,00 kr
Log in for availability
1 * 1 mg
Product Details & Documents

AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.

  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal

 : For research use only.

  • Storage Temperature:Fryser
Specifications

Specifications

  • Pakn:
    1 mg

Specifications