Orders LinkContactUsLinkComponent
Stand Rods, without Thread
Stand Rods, without Thread
13 mm, 300 mm
Katalog # BOCH5180-300
Leverandør:   BOCHEM
Stand Rods, without Thread
Katalog # BOCH5180-300
Leverandør:  BOCHEM
Stand Rods, without Thread
13 mm, 300 mm
Price
198,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel, 18/10 steel.
  • Stand rods for stand bases
  • Without thread
Specifikationer
  • Ø:
    13 mm
  • Længde:
    300 mm
  • Seq:
    02