Orders LinkContactUsLinkComponent
Polypropylene Glycol Standards
Polypropylene Glycol Standards
Polypropylene glycol standard, molecular weight: 450 Da, 1 g, 1
Katalog # AGLSPSS-PPG450
Leverandør:   AGILENT
Polypropylene Glycol Standards
Katalog # AGLSPSS-PPG450
Leverandør:  AGILENT
CAS nummer:  
Polypropylene Glycol Standards
Polypropylene glycol standard, molecular weight: 450 Da, 1 g, 1
CAS nummer:  
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Polypropylene glycol standard, PDI <1,15, 1 g.
Specifikationer
Specifikationer
  • Pakn:
    1
  • Beskrivelse:
    Polypropylene glycol standard, molecular weight: 450 Da, 1 g
Specifikationer