Polypropylene Glycol Standards
Polypropylene glycol standard, molecular weight: 4500 Da, 1 g, 1
Katalog # AGLSPSS-PPG4.5K
Leverandør: AGILENT
Gå til:
Product Details & Documents
Polypropylene glycol standard, PDI <1,15, 1 g.
Specifikationer
Specifikationer
- Pakn:1
- Beskrivelse:Polypropylene glycol standard, molecular weight: 4500 Da, 1 g