Calibrated Disposable Plastic Inoculation Loops
Rigid (hard) plastic inoculation loop, 10 µl, mørkeblå, 40/pack
Supplier: Copan
Gå til:
Product Details & Documents
COPAN calibrated plastic inoculation loops have an ultra-smooth surface to avoid gouging agar when planting and streaking.
Loops are free of lubricants, oils, and electrostatic charges.
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Pakket:40/pack
- Kapacitet:10 µl
- Farve:mørkeblå
- Beskrivelse:Rigid (hard) plastic inoculation loop
- Seq:14