Calibrated Disposable Plastic Inoculation Loops
Rigid (hard) plastic inoculation loop, 1 µl, mørkegrøn, 40/pack
Katalog # COPA8175CSR40H
Leverandør: Copan
Gå til:
Product Details & Documents
COPAN calibrated plastic inoculation loops have an ultra-smooth surface to avoid gouging agar when planting and streaking.
Loops are free of lubricants, oils, and electrostatic charges.
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Pakket:40/pack
- Kapacitet:1 µl
- Farve:mørkegrøn
- Beskrivelse:Rigid (hard) plastic inoculation loop
- Seq:12