Orders LinkContactUsLinkComponent
Calibrated Disposable Plastic Inoculation Loops
Calibrated Disposable Plastic Inoculation Loops
Rigid (hard) plastic inoculation loop, 1 µl, mørkegrøn, 40/pack
Katalog # COPA8175CSR40H
Leverandør:   Copan
Calibrated Disposable Plastic Inoculation Loops
Katalog # COPA8175CSR40H
Leverandør:  Copan
Calibrated Disposable Plastic Inoculation Loops
Rigid (hard) plastic inoculation loop, 1 µl, mørkegrøn, 40/pack
Price
903,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents
COPAN calibrated plastic inoculation loops have an ultra-smooth surface to avoid gouging agar when planting and streaking.
Loops are free of lubricants, oils, and electrostatic charges.
Documents
Specifikationer
  • Pakket:
    40/pack
  • Kapacitet:
    1 µl
  • Farve:
    mørkegrøn
  • Beskrivelse:
    Rigid (hard) plastic inoculation loop
  • Seq:
    12