Orders LinkContactUsLinkComponent
Calibrated Disposable Plastic Inoculation Loops
Calibrated Disposable Plastic Inoculation Loops
Flexible (soft) plastic inoculation loop, 10 µl, lyseblå, 40/pack
Katalog # COPA179CSR40
Leverandør:   Copan
Calibrated Disposable Plastic Inoculation Loops
Katalog # COPA179CSR40
Leverandør:  Copan
Calibrated Disposable Plastic Inoculation Loops
Flexible (soft) plastic inoculation loop, 10 µl, lyseblå, 40/pack
Price
903,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents
COPAN calibrated plastic inoculation loops have an ultra-smooth surface to avoid gouging agar when planting and streaking.
Loops are free of lubricants, oils, and electrostatic charges.
Documents
Specifikationer
  • Pakket:
    40/pack
  • Kapacitet:
    10 µl
  • Farve:
    lyseblå
  • Beskrivelse:
    Flexible (soft) plastic inoculation loop
  • Seq:
    13