Orders LinkContactUsLinkComponent
Calibrated Disposable Plastic Inoculation Loops
Inoculation needle with 1,45 mm Ø, 200 mm length, −, Violet, 20/pouch
  COPA176CS20
 :   Copan
Calibrated Disposable Plastic Inoculation Loops
  COPA176CS20
 :  Copan
Calibrated Disposable Plastic Inoculation Loops
Inoculation needle with 1,45 mm Ø, 200 mm length, −, Violet, 20/pouch
627,00 kr
1 * 1.000 Items
COPAN calibrated plastic inoculation loops have an ultra-smooth surface to avoid gouging agar when planting and streaking.
Loops are free of lubricants, oils, and electrostatic charges.
  • Pakket:
    20/pouch
  • Kapacitet:
  • Farve:
    Violet
  • Beskrivelse:
    Inoculation needle with 1,45 mm Ø, 200 mm length
  • Seq:
    01