[EN]11 MULTI-ELEMENT STNDRD SOLUTION IN 1 * 500 mL
Katalog # CMPLRM098086L1.2
Leverandør: CUSTOM MADE CHEMICALS LAB
[EN]11 MULTI-ELEMENT STNDRD SOLUTION IN 1 * 500 mL
Katalog # CMPLRM098086L1.2
Leverandør: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Leverandør: Merck
VWR® AVS TITRINORM, bufferopløsninger, 20 °C
Leverandør: VWR Chemicals
VWR®, Refractive index Certified Reference Material at 20, 25 and 30 °C
Leverandør: VWR Chemicals
Multi-element kvalitetskontrolstandard 100 mg/L, 28 elementer i salpetersyre 5% ARISTAR® til ICP
Leverandør: VWR Chemicals
Buffer solutions as per DIN 19266-01: 2000
Leverandør: WTW
Kaliumdihydrogenorthophosphat 0.2 mol/l in aqueous solution Reag. Ph. Eur. 1069601
Leverandør: VWR Chemicals
Ethyl Acetate, Reference Standard
Leverandør: USP
2-HEXANONE 1 * 1 mL
Leverandør: Merck
Reagents for alkalinity determination using Lovibond® MD photometers
Leverandør: Lovibond
Buffer Solutions
Leverandør: Thermo Fisher Scientific
Turbidity standards, Stablcal®
Leverandør: Hach