Accessories for SCAT Level Controls
Holder for disc sensor for safety waste cap S60/61, three levels adjustable, PE_HD
Katalog # SCTV106731
Leverandør: S.C.A.T.
Gå til:
Product Details & Documents
Necessary and helpful accessories for use with SCAT electronic level controls.
- Switching cables
- Disk sensors
- Signal cables
- Power supply units
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Specifikationer
- Beskrivelse:Holder for disc sensor for safety waste cap S60/61, three levels adjustable, PE_HD
- Seq:2



