Forceps, pointed
Straight, 115 mm
Katalog # COMC200-0115
Leverandør: COMACO HANDEL
Gå til:
Product Details & Documents
Stainless steel, grade 18-8.
- Pointed
- With serrated grip and tips
Specifikationer
- Model:Straight
- Længde:115 mm
- Seq:02