Plasmid DNA buffers
DNA wash buffer concentrate, low salt spin column was solution (makes 250 ml), 250 mL
Leverandør: OMEGA BIO-TEK
Gå til:
Product Details & Documents
Documents
Specifikationer
Specifikationer
- Pakn:250 mL
- Beskrivelse:DNA wash buffer concentrate, low salt spin column was solution (makes 250 ml)