Krytech 643 Cut Resistant Gloves
Product Details & Documents
KryTech 643 is comfort, suppleness and breathability without any compromise on cut protection and durability can be used with touch devices and touch screen.
- A seamless plaited glove for a maximum comfort, dexterity and flexibility while providing optimal cut protection
- Extended use guaranteed by our exclusive process RESICOMFORT
- Free of harmful substances: OEKO-TEX® standard 100, DMF free
- Interior finish seamless textile support from HDPE fibres
- Exterior finish reinforced grip
- Dermatologically approved
- Gauge: 15
It can be used with touch devices and touch screen. Silicone free to prevent traces, defects on metal sheet and glass prior to painting. Washable one time at 40 °C without any alteration on performances*
* Tests carried out on unworn gloves which differs from the reality in the field.
Certifications: ISO certified. This product is not classified hazardous according to the regulation (EC) n°1272/2008 of the European Parliament and of the council. This product does not contain more than 0.1 % of substance of very high concern (SVHC) or any substance included in the annex XVII of the regulation n° 1907/2006 of the European Parliament and of the Council (REACH). UE type certificate or CE type examination certificate: 0075/014/162/10/20/2586. Issued by the notified body nr: 0075 CTC - 4, rue Hermann Frenkel - 69367 Lyon. Cedex 07- FRANCE.
Specifikationer
- Handske størrelse:6
- Pakket:12 pair/bag
- Farve:sort/grå
- Længde:230 - 280 mm
- Seq:01
- Model:Krytech 643
- Cat. No.:MAPA34643016
- Ambidextrous/Hånd-specifik:Hånd-specifik
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



