Bottles, Narrow Neck, Round
100 ml, GL 18, 37 mm, 114 mm, 18 mm
Katalog # 215-4658
Leverandør: BERLIN PACKAGING RIXIUS GMBH
Gå til:
Product Details & Documents
Clear or amber glass without screw cap.
- Good chemical and physical properties
- For applications that do not involve high thermal stresses
- Round thread in accordance with DIN 168
Excellent for sampling, storing and transporting liquids. Amber coloured to protect light-sensitive materials.
Documents
RIXI10501001018ADO_TDS_EN
Datablad
Specifikationer
- Udv.Ø:37 mm
- Gevind:GL 18
- Kapacitet:100 ml
- Højde:114 mm
- Seq:14
- Hals indv. Ø:18 mm