Bottles, Narrow Neck, Round
30 ml, GL 18, 37 mm, 83,8 mm, 18 mm
215-4657
Clear or amber glass without screw cap.
- Good chemical and physical properties
- For applications that do not involve high thermal stresses
- Round thread in accordance with DIN 168
Excellent for sampling, storing and transporting liquids. Amber coloured to protect light-sensitive materials.
- Udv.Ø:37 mm
- Gevind:GL 18
- Kapacitet:30 ml
- Højde:83,8 mm
- Seq:11
- Hals indv. Ø:18 mm