Orders LinkContactUsLinkComponent
VHB DNA Washing Buffer
VHB DNA Washing Buffer
VHB buffer, 440 mL
Katalog # OMEGVHB-440
Leverandør:   OMEGA BIO-TEK
VHB DNA Washing Buffer
Katalog # OMEGVHB-440
Leverandør:  OMEGA BIO-TEK
CAS nummer:  
VHB DNA Washing Buffer
VHB buffer, 440 mL
CAS nummer:  
Price
4.690,00 kr
Log ind for tilgængelighed
1 * 440 mL
Product Details & Documents
VHB buffer must be diluted with ethanol before use.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Specifikationer
Specifikationer
  • Pakn:
    440 mL
  • Beskrivelse:
    VHB buffer