VHB DNA Washing Buffer
VHB buffer, 440 mL
Katalog # OMEGVHB-440
Leverandør: OMEGA BIO-TEK
Gå til:
Product Details & Documents
VHB buffer must be diluted with ethanol before use.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Specifikationer
Specifikationer
- Pakn:440 mL
- Beskrivelse:VHB buffer