Orders LinkContactUsLinkComponent
Cryoboxes, Cardboard
Cryoboxes, Cardboard
9×9, Green/pink, 52×134×134 mm
  TENATE22878
 :   TENAK
Cryoboxes, Cardboard
  TENATE22878
 :  TENAK
Cryoboxes, Cardboard
Cryoboxes, Cardboard
9×9, Green/pink, 52×134×134 mm
668,00 kr
1 * 36 Items
Colored cryoboxes possible to orient in different ways for classic colour coding, tracking work in progress, identifying the owner of the box, and monitoring sample thawing.
  • PP coated water-repellent surface
  • Internal printed alpha numeric grid
For space-saving solutions, the cryobox is also available in a convenient flat-packed Origami box format.
  • Rum:
    9×9
  • B×D×H udv.:
    52×134×134 mm
  • Farve:
    Green/pink
  • Seq:
    03