Cryoboxes, Cardboard
9×9, Green/pink, 52×134×134 mm
TENATE22878
: TENAK
Colored cryoboxes possible to orient in different ways for classic colour coding, tracking work in progress, identifying the owner of the box, and monitoring sample thawing.
- PP coated water-repellent surface
- Internal printed alpha numeric grid
For space-saving solutions, the cryobox is also available in a convenient flat-packed Origami box format.
- Rum:9×9
- B×D×H udv.:52×134×134 mm
- Farve:Green/pink
- Seq:03