Orders LinkContactUsLinkComponent
Cryoboxes, Cardboard
Cryoboxes, Cardboard
10×10, blå/orange, 52×134×134 mm
Catalog # TENATE22875
Supplier:   TENAK
Cryoboxes, Cardboard
Catalog # TENATE22875
Supplier:  TENAK
Cryoboxes, Cardboard
Cryoboxes, Cardboard
10×10, blå/orange, 52×134×134 mm
Price
668,00 kr
Log in for availability
1 * 36 Items
Product Details & Documents
Colored cryoboxes possible to orient in different ways for classic colour coding, tracking work in progress, identifying the owner of the box, and monitoring sample thawing.
  • PP coated water-repellent surface
  • Internal printed alpha numeric grid
For space-saving solutions, the cryobox is also available in a convenient flat-packed Origami box format.
Specifications
  • Rum:
    10×10
  • B×D×H udv.:
    52×134×134 mm
  • Farve:
    blå/orange
  • Seq:
    05