Cryoboxes, Cardboard
10×10, Yellow/purple, 52×134×134 mm
Katalog # TENATE22871
Leverandør: TENAK
Gå til:
Product Details & Documents
Colored cryoboxes possible to orient in different ways for classic colour coding, tracking work in progress, identifying the owner of the box, and monitoring sample thawing.
- PP coated water-repellent surface
- Internal printed alpha numeric grid
For space-saving solutions, the cryobox is also available in a convenient flat-packed Origami box format.
Specifikationer
- Rum:10×10
- B×D×H udv.:52×134×134 mm
- Farve:Yellow/purple
- Seq:04