Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Vial kits

Vial kits

1,50 ml, PP, klar, Screw cap, 9 mm, Clear PP, 9 mm
Catalog # HYPE6EKSV9PP-15
Vial kits
Catalog # HYPE6EKSV9PP-15
Vial kits

1,50 ml, PP, klar, Screw cap, 9 mm, Clear PP, 9 mm

Price
517,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents
Specifications
  • Cap størrelse:
    9 mm
  • Septum:
    Clear PP, 9 mm
  • Kapacitet:
    1,50 ml
  • Farve:
    klar
  • Materiale:
    PP
  • Låg type:
    Screw cap