Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Vial kits

0,80 ml, PE, klar, Snap cap, 11 mm, Blue PE, 11 mm
  HYPE6EKRV11PP-08
Vial kits
  HYPE6EKRV11PP-08
Vial kits

0,80 ml, PE, klar, Snap cap, 11 mm, Blue PE, 11 mm

 
366,00 kr
   
1 * 100 Items
 
 
  • Cap størrelse:
    11 mm
  • Septum:
    Blue PE, 11 mm
  • Kapacitet:
    0,80 ml
  • Farve:
    klar
  • Materiale:
    PE
  • Låg type:
    Snap cap
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.