Dunke
2 L, UN approval: UN 3H1/Y/1.4, 115×155×175 mm, GL 45
KART1780
: KARTELL
HDPE, translucent with black screw cap.
- With comfortable handle and secure seal cap following DIN recommendations
- Conform to UN regulation on transportation of dangerous liquids
- Suitable for foodstuff
Ideal for storing and transporting liquids.
- Gevind:GL 45
- Kapacitet:2 L
- Beskrivelse:UN approval: UN 3H1/Y/1.4
- B×D×H:115×155×175 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



