0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
ContactUsLinkComponent
[EN]MIXTURE 26 COMPONENTS 1 * 100 mL
CMPL0D7BK15NTRF.L1
:
CUSTOM MADE CHEMICALS LAB
[EN]MIXTURE 26 COMPONENTS 1 * 100 mL
CMPL0D7BK15NTRF.L1
:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
: Merck
Nitrat 1.000 mg/l i vand (af NaNO<sub>3</sub>) Certipur®, Supelco®
: Merck
Kaliumhydrogenphthalat, Certipur® Reag. Ph. Eur., Reag. USP volumetric standard, secondary reference material for alkalimetry, Supelco®
: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
: Merck
Kaliumchlorid 3 mol/l (3 N) in aqueous solution, Supelco®
: Merck
VWR® AVS TITRINORM, bufferopløsninger, 20 °C
: VWR Chemicals
Phosphat 1000 mg/l i vand (af KH<sub>2</sub>PO<sub>4</sub>) Certipur®, Supelco®
: Merck
Buffer solutions in sachets, Certipur®, Supelco®
: Merck
VWR®, Conductivity standard solutions
: VWR Chemicals
pH buffers, ready to use
: WTW
pH Standard Solutions, IUPAC, Certified
: Hach