Labels for BBP33 printers, type B-499
Printer labels, B-499, Nylon, Permanent akryl, hvid, BBP33, 12,7×36,4 mm, 2500/roll
BRDY153090
: Brady
These nylon cloth labels are used for electrical and automation identification, inside-panel component labelling, wire and cable marking.
- Printer:BBP33
- Model:B-499
- Farve:hvid
- Beskrivelse:Printer labels
- Dimensioner:12,7×36,4 mm
- Materiale:Nylon
- Seq:08
- Pakket:2500/roll
- Klæbestof:Permanent akryl