Orders LinkContactUsLinkComponent
Labels for BBP33 printers, type B-499
Labels for BBP33 printers, type B-499
Printer labels, B-499, Nylon, Permanent akryl, hvid, BBP33, 25,4×36,4 mm, 2300/roll
Katalog # BRDY153092
Leverandør:   Brady
Labels for BBP33 printers, type B-499
Katalog # BRDY153092
Leverandør:  Brady
Labels for BBP33 printers, type B-499
Labels for BBP33 printers, type B-499
Printer labels, B-499, Nylon, Permanent akryl, hvid, BBP33, 25,4×36,4 mm, 2300/roll
Price
3.980,00 kr
Log ind for tilgængelighed
1 * 1 ROL
Product Details & Documents
These nylon cloth labels are used for electrical and automation identification, inside-panel component labelling, wire and cable marking.
Specifikationer
  • Printer:
    BBP33
  • Model:
    B-499
  • Farve:
    hvid
  • Beskrivelse:
    Printer labels
  • Dimensioner:
    25,4×36,4 mm
  • Materiale:
    Nylon
  • Seq:
    22
  • Pakket:
    2300/roll
  • Klæbestof:
    Permanent akryl