Orders LinkContactUsLinkComponent
Labels for BBP33 printers, type B-499
Labels for BBP33 printers, type B-499
Printer labels, B-499, Nylon, Permanent akryl, hvid, BBP33, 19,0×19,0 mm, 1000/roll
  BRDY153088
 :   Brady
Labels for BBP33 printers, type B-499
  BRDY153088
 :  Brady
Labels for BBP33 printers, type B-499
Labels for BBP33 printers, type B-499
Printer labels, B-499, Nylon, Permanent akryl, hvid, BBP33, 19,0×19,0 mm, 1000/roll
1.500,00 kr
1 * 1 ROL
These nylon cloth labels are used for electrical and automation identification, inside-panel component labelling, wire and cable marking.
  • Printer:
    BBP33
  • Model:
    B-499
  • Farve:
    hvid
  • Beskrivelse:
    Printer labels
  • Dimensioner:
    19,0×19,0 mm
  • Materiale:
    Nylon
  • Seq:
    11
  • Pakket:
    1000/roll
  • Klæbestof:
    Permanent akryl