Cerex® TRACE-J SPE Columns
Column Cerex TRACE-J, 15 µm, 3 ml, 35 mg
Katalog # TCAT709-335C
Leverandør: Tecan
Gå til:
Product Details & Documents
Strong cationic columns with small particle and pore sizes for aggressive cleanup of basic compounds.
- Ion-exchange
- Liquid handler compatible
A 18 µm polymer micro particulate with strong cationic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Ideal for basic target compounds. Formulated for pain panel/FNT.
Specifikationer
- Partikelstørrelse:15 µm
- Kapacitet:3 ml
- Beskrivelse:Column Cerex TRACE-J
- Seq:01
- Lejevægt:35 mg