Cerex® THC SPE Columns
Cerex Atlas® column THC, THC, 80 Å, 17 µm, 6 ml, 65 mg
Katalog # TCAT682-0506C
Leverandør: Tecan
Gå til:
Product Details & Documents
Polymer micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample preparation. Specially formulated for THC/THCA testing.
Specifikationer
- Partikelstørrelse:17 µm
- Porestørrelse:80 Å
- Kapacitet:6 ml
- Beskrivelse:Cerex Atlas® column THC
- Seq:04
- Lejevægt:65 mg
- Fase:THC