Cerex® PSAXS SPE Columns
Column Cerex PSAXS, PSAX-S, 300 Å, 10 µm, 3 ml, 50 mg
Katalog # TCAT606-0503C
Leverandør: Tecan
Gå til:
Product Details & Documents
Mixed mode reverse-phase with an anion exchanger for acidic compounds.
- Ion-exchange
- Liquid handler compatible
A 10 µm silica micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep.
Specifikationer
- Partikelstørrelse:10 µm
- Porestørrelse:300 Å
- Kapacitet:3 ml
- Beskrivelse:Column Cerex PSAXS
- Seq:02
- Lejevægt:50 mg
- Fase:PSAX-S