Cerex® HPSCX SPE Columns
Column Cerex HPSCX, HPSCX, 30 µm, 3 ml, 35 mg
Catalog # TCAT427-0353C
Supplier: Tecan
Jump to:
Product Details & Documents
Mixed mode sorbent with both reverse-phase and strong cationic exchange chemistries.
- Ion-exchange
- Liquid handler compatible
Its high surface area provides excellent capacity and a high selectivity for basic analytes. Ideal for drug monitoring, screening, identification, confirmation and quantitation.
Specifications
- Partikelstørrelse:30 µm
- Kapacitet:3 ml
- Beskrivelse:Column Cerex HPSCX
- Seq:01
- Lejevægt:35 mg
- Fase:HPSCX