Cerex® WWP2 SPE Columns
Column Cerex WWP2, WWP2, 300 Å, 30 µm, 1 ml, 10 mg
Katalog # TCAT417-0101R
Leverandør: Tecan
Gå til:
Product Details & Documents
Polymeric water-wettable resin ideal for reverse-phase retention of aqueous samples.
- Reversed-phase
- Liquid handler compatible
Special water wettable polymer with 300 Å pore size ideal for protein digest clean up and other protein related workflows.
Specifikationer
- Partikelstørrelse:30 µm
- Porestørrelse:300 Å
- Kapacitet:1 ml
- Beskrivelse:Column Cerex WWP2
- Seq:01
- Lejevægt:10 mg
- Fase:WWP2