Cerex® WWP SPE Columns
Cerex Atlas® column WWP, WWP, 80 Å, 20 µm, 1 ml, 20 mg
Katalog # TCAT676-0201R
Leverandør: Tecan
Gå til:
Product Details & Documents
Microparticulate water wettable polymer provides great chemical stability and ideal for synthetic cannabinoids analysis.
Specifikationer
- Partikelstørrelse:20 µm
- Porestørrelse:80 Å
- Kapacitet:1 ml
- Beskrivelse:Cerex Atlas® column WWP
- Seq:01
- Lejevægt:20 mg
- Fase:WWP