Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Cerex® HPSAX SPE Columns

Column Cerex HPSAX, 20 µm, 1 ml, 40 mg
Katalog # TCAT412-0401R
Leverandør:   Tecan
Cerex® HPSAX SPE Columns
Katalog # TCAT412-0401R
Leverandør:  Tecan
Cerex® HPSAX SPE Columns

Column Cerex HPSAX, 20 µm, 1 ml, 40 mg

Price
Spørg for pris
Log ind for tilgængelighed
1 * 96 Items
Product Details & Documents

Polymeric strong anionic exchange chemistry.

  • Ion-exchange
  • Liquid handler compatible

20 µm polymer micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in fast and uniform sample prep. Ideal for acidic compounds.

Specifikationer
  • Partikelstørrelse:
    20 µm
  • Kapacitet:
    1 ml
  • Beskrivelse:
    Column Cerex HPSAX
  • Seq:
    05
  • Lejevægt:
    40 mg