Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Cerex® HPSAX SPE Columns

Column Cerex HPSAX, 20 µm, 1 ml, 20 mg
  TCAT412-0201R
 :   Tecan
Cerex® HPSAX SPE Columns
  TCAT412-0201R
 :  Tecan
Cerex® HPSAX SPE Columns

Column Cerex HPSAX, 20 µm, 1 ml, 20 mg

 
 
   
1 * 96 Items
 

Polymeric strong anionic exchange chemistry.

  • Ion-exchange
  • Liquid handler compatible

20 µm polymer micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in fast and uniform sample prep. Ideal for acidic compounds.

 
  • Partikelstørrelse:
    20 µm
  • Kapacitet:
    1 ml
  • Beskrivelse:
    Column Cerex HPSAX
  • Seq:
    03
  • Lejevægt:
    20 mg