Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cerex® HPSAX SPE Columns

Cerex® HPSAX SPE Columns

Column Cerex HPSAX, 20 µm, 1 ml, 10 mg
Katalog # TCAT412-0101R
Leverandør:   Tecan
Cerex® HPSAX SPE Columns
Katalog # TCAT412-0101R
Leverandør:  Tecan
Cerex® HPSAX SPE Columns

Column Cerex HPSAX, 20 µm, 1 ml, 10 mg

Price
Spørg for pris
Log ind for tilgængelighed
1 * 96 Items
Product Details & Documents

Polymeric strong anionic exchange chemistry.

  • Ion-exchange
  • Liquid handler compatible

20 µm polymer micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in fast and uniform sample prep. Ideal for acidic compounds.

Specifikationer
  • Partikelstørrelse:
    20 µm
  • Kapacitet:
    1 ml
  • Beskrivelse:
    Column Cerex HPSAX
  • Seq:
    02
  • Lejevægt:
    10 mg
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.