Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cerex® C18 SPE Heavy Load Columns

Cerex® C18 SPE Heavy Load Columns

Column C18 Heavy load, 1 ml, 20 mg
Katalog # TCAT618-0201R
Leverandør:   Tecan
Cerex® C18 SPE Heavy Load Columns
Katalog # TCAT618-0201R
Leverandør:  Tecan
Cerex® C18 SPE Heavy Load Columns

Column C18 Heavy load, 1 ml, 20 mg

Price
Spørg for pris
Log ind for tilgængelighed
1 * 96 Items
Product Details & Documents

Endcapped C18 chemistry.

  • Reversed-phase
  • Liquid handler compatible

A 10 µm micro particulate with endcapped C18 chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample preparation.

Specifikationer
  • Kapacitet:
    1 ml
  • Beskrivelse:
    Column C18 Heavy load
  • Seq:
    01
  • Lejevægt:
    20 mg
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.