Orders LinkContactUsLinkComponent
Cerex® C18 SPE Heavy Load Columns
Cerex® C18 SPE Heavy Load Columns
Column C18 NON-EC, 3 ml, 100 mg
Katalog # TCAT602-1003
Leverandør:   Tecan
Cerex® C18 SPE Heavy Load Columns
Katalog # TCAT602-1003
Leverandør:  Tecan
Cerex® C18 SPE Heavy Load Columns
Column C18 NON-EC, 3 ml, 100 mg
Price
Spørg for pris
Log ind for tilgængelighed
1 * 50 Items
Product Details & Documents
Endcapped C18 chemistry.
  • Reversed-phase
  • Liquid handler compatible
A 10 µm micro particulate with endcapped C18 chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample preparation.
Specifikationer
  • Kapacitet:
    3 ml
  • Beskrivelse:
    Column C18 NON-EC
  • Lejevægt:
    100 mg