Resolvex® M10 Manual Positive Pressure Processor
Resolvex® M10 48 instrument, 355×305×390 mm
Katalog # TCAT289-0004
Leverandør: Tecan
Gå til:
Product Details & Documents
Benchtop manual positive pressure processor.
- Individual flow restrictors ensure parallel processing even when some wells are vacant
- Only a gas supply is needed for operation
- Dual-level gas delivery system offers fine controls of flow and pressure
Perfect for entry into positive pressure processing and workflows, the intuitive design of the Resolvex® M10 makes it easy to use with minimal training required.
Specifikationer
- Beskrivelse:Resolvex® M10 48 instrument, 355×305×390 mm
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- SPE column rack for 6 ml columns for 48 place, aluminum, W×D×H: 10,8×3,6×3,5"SPE column rack for 6 ml columns for 48 place, aluminum, W×D×H: 10,8×3,6×3,5"TCAT279-0023Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- SPE column rack for 1 ml cerex columns, aluminum, W×D×H: 10,8×3,6×3,1"SPE column rack for 1 ml cerex columns, aluminum, W×D×H: 10,8×3,6×3,1"TCAT279-0021Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- SPE column rack for 3 ml columns for 48 place, aluminum, W×D×H: 10,8×3,6×3,5"SPE column rack for 3 ml columns for 48 place, aluminum, W×D×H: 10,8×3,6×3,5"TCAT279-0022Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Partial plate holder for 1 ml columns (generic), aluminum, W×D×H: 3,1×3,4×5,0"Partial plate holder for 1 ml columns (generic), aluminum, W×D×H: 3,1×3,4×5,0"TCAT253-0006Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Waste barrier insert (long), plastic, W×D×H: 4,4×3,0×1,6"Waste barrier insert (long), plastic, W×D×H: 4,4×3,0×1,6"TCAT253-0036-SPLog ind for tilgængelighed1 * 5 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Collection rack for 12×75 mm tubes for 48 place system, stainless steel, W×D×H: 10,7×3,5×3,8"Collection rack for 12×75 mm tubes for 48 place system, stainless steel, W×D×H: 10,7×3,5×3,8"TCAT279-0015Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Partial plate holder for NBE columns (generic), aluminum, W×D×H: 3,1×3,4×5,0"Partial plate holder for NBE columns (generic), aluminum, W×D×H: 3,1×3,4×5,0"TCAT253-0007Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Collection rack for 16×100 tubes for 48 mm place system, stainless steel, W×D×H: 10,7×3,5×3,8"Collection rack for 16×100 tubes for 48 mm place system, stainless steel, W×D×H: 10,7×3,5×3,8"TCAT279-0016Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Collection rack for 12×32 mm autosampler vials for 48 place systems, stainless steel, W×D×H: 10,7×3,5×3,8"Collection rack for 12×32 mm autosampler vials for 48 place systems, stainless steel, W×D×H: 10,7×3,5×3,8"TCAT279-0017Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Mat Luer seal pull tab silicone, H=5,11 mm
Produktinformation
- Katalog #TilgængelighedPrice
- Collection rack to fit 12×32 mm autosampler vials, stainless steel, W×D×H: 10,7×3,5×3,9"Collection rack to fit 12×32 mm autosampler vials, stainless steel, W×D×H: 10,7×3,5×3,9"TCAT279-0018Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Reservoir 96 well 300 ml pyramid, plastic, W×D×H: 5,0×3,4×1,7"Reservoir 96 well 300 ml pyramid, plastic, W×D×H: 5,0×3,4×1,7"TCAT700-4009Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Mat 96 well pre-slit silic BlankSilicone, H=5,11 mmMat 96 well pre-slit silic BlankSilicone, H=5,11 mmTCAT278-0060Log ind for tilgængelighed1 * 10 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Plate V-bottom solid 5 ml, plastic, W×D×H: 5,0×3,4×0,6"Plate V-bottom solid 5 ml, plastic, W×D×H: 5,0×3,4×0,6"TCAT700-4008Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Rack sample 6CC 20 mm spacing, plastic, W×D×H: 5,0×3,4×2,9"Rack sample 6CC 20 mm spacing, plastic, W×D×H: 5,0×3,4×2,9"TCAT30219826Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Rack sample 3CC 20 mm spacing, plastic, W×D×H: 5,0×3,4×2,9"Rack sample 3CC 20 mm spacing, plastic, W×D×H: 5,0×3,4×2,9"TCAT30219827Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Rack collection tube 12×75 mm assay, plastic, W×D×H: 5,0×3,6×3,2"Rack collection tube 12×75 mm assay, plastic, W×D×H: 5,0×3,6×3,2"TCAT30219825Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Rack vial autosampler assay, plastic, W×D×H: 5,3×3,6×1,7"Rack vial autosampler assay, plastic, W×D×H: 5,3×3,6×1,7"TCAT30219821Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Rack MN giga assay, plastic, W×D×H: 5,0×3,4×4,4"Rack MN giga assay, plastic, W×D×H: 5,0×3,4×4,4"TCAT30219819Log ind for tilgængelighed1 * 1 Items
Produktinformation
- Katalog #TilgængelighedPrice
- Rack QIAGEN-tip mega assay, plastic, W×D×H: 5,0×3,4×4,3"Rack QIAGEN-tip mega assay, plastic, W×D×H: 5,0×3,4×4,3"TCAT30219818Log ind for tilgængelighed1 * 1 Items
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



