Orders LinkContactUsLinkComponent
Cover glasses
High precision cover glasses, 85×127 mm, #1,5, Borosilikatglas
  MARI01079990935
 :   MARIENFELD
Cover glasses
  MARI01079990935
Cover glasses
Cover glasses
High precision cover glasses, 85×127 mm, #1,5, Borosilikatglas
1 * 1.000 Items
Cover slides suitable for fluorescence microscopy.
  • Borosilicate glass D 263™ M of hydrolytic class 1
Recommended for objectives with high numerical aperture and resolution.
  • Antal:
    #1,5
  • Beskrivelse:
    High precision cover glasses
  • L×B:
    85×127 mm
  • Materiale:
    Borosilikatglas
  • Seq:
    05