Printer Accessories for Models M511 and M211
Product Details & Documents
Accessories for Brady Printer Models M511 and M211.
- M511 hard case - transport and store your M511 label printer
- M511 magnet - magnet attaches the M511 printer to panels, I-beams and other magnetic-receptive surfaces
- M511 utility hook - hang printer easily on a cart, tool belt, rack or bucket organiser
- M511 and M211 power brick - compatible with all mobile devices, 70-minute charge time
- M511 and M211 AC adapter - to power and charge M511 and M211 printers with power supply voltage: 110 to 240 V
Hard case - Protects your printer from impacts and dust. Keeps printer, accessories and label cartridges organised and on hand. Durable hard case allows for easy mobility.
Magnet - Get hands-free operation by affixing to a metal surface. Attaches to back of printer with screws to ensure stability.
Utility hook - Attaches to back of printer with screws to ensure stability. Made from heavy-duty steel for added durability.
Power brick - A fully-charged 10000mAh Power Brick can charge your M511 or M211 printer up to three times. LED digital display shows the percentage of power remaining. Outputs include: USB-C, USB-A1/A2.
AC adapter - USB A-to-C dongle allows you to charge the printer using a USB port in a vehicle or on a laptop, plus can be used to connect the M511 to Brady workstation label creation software.
Bestillingsoplysninger: Power brick (BRDY176499) has a rechargeable lithium-ion battery, must ship ground.
Specifikationer
- Beskrivelse:Battery, rechargeable lithium-ion, includes (1) power brick (power brick [BRADY176499] has a rechargeable lithium-ion battery, must ship ground), 50,8W×23,8D×115,8H mm
- Seq:02
- Til:M511; M211