[EN]MIX RD10002810.10 10X10ML AMPOULE 1 * 10 Items
Katalog # CMPLRD10002810.10
Leverandør: CUSTOM MADE CHEMICALS LAB
[EN]MIX RD10002810.10 10X10ML AMPOULE 1 * 10 Items
Katalog # CMPLRD10002810.10
Leverandør: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Paraffin, Solidification point: 54 - 56 °C, Q Path® for microscopy
Leverandør: VWR Chemicals
Natriumhydroxid 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Leverandør: VWR Chemicals
Papir-pH-indikatorer, MQuant®, Supelco®
Leverandør: Merck
Visuelle farvekits, MColortest™, Supelco®
Leverandør: Merck
Immersion oil for microscopy, Sigma-Aldrich®
Leverandør: Merck
Natriumhydroxid, piller, EMSURE® for analysis, Supelco®
Leverandør: Merck
VWR®, Immersion Oil
Leverandør: VWR Collection
pH-teststrimler, MQuant®, Supelco®
Leverandør: Merck
Natriumhydroxid, piller, ren, Supelco®
Leverandør: Merck
Natriumhydroxid 32% in aqueous solution, AnalaR NORMAPUR® analytical reagent, for nitrogen analysis
Leverandør: VWR Chemicals
Ammoniakvand 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
Leverandør: Merck
VWR®, OCT Embedding matrix for frozen sections
Leverandør: VWR Chemicals
Formalin 10%, Q Path®, buffered
Leverandør: VWR Chemicals
Teststrips, MQuant™, Supelco®
Leverandør: Merck
Teststrips, MQuant™, Supelco®
Leverandør: Merck
Test kits, photometric, COD, LCK cuvette tests
Leverandør: Hach