Orders LinkContactUsLinkComponent
Accessories for label printer, M611
Accessories for label printer, M611
Magnet, black, 111.25 mm, M611 label printer
Katalog # BRDY175604
Leverandør:   Brady
Accessories for label printer, M611
Katalog # BRDY175604
Leverandør:  Brady
Accessories for label printer, M611
Accessories for label printer, M611
Magnet, black, 111.25 mm, M611 label printer
Price
489,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Various accessories for the M611 label printer.
  • Hard case and magnet are available
Hard case for M611 label printer – Designed to carry your M611 label printer and extra supplies – anywhere you need it. Durable hard case protects your printer against impacts and dust while allowing for easy mobility. Keeps printer accessories organised and on hand. Protect your portable M611 label printer and keep accessories organised with this durable plastic carrying case.
M611 magnet – Get hands-free operation by affixing to panel door or metal surface. Attaches to back of printer with screws to ensure stability. Designed for use with the M611 Label Printer. Use your M611 Label Printer just about anywhere. This magnet attaches the M611 printer to panels, I-beams and other magnetic-receptive surfaces during use or storage.
Specifikationer
  • Beskrivelse:
    Magnet, black, 111.25 mm
  • Til:
    M611 label printer