Orders LinkContactUsLinkComponent
Cone folded filter papers, Whatman™
Cone folded filter papers, Whatman™
2, 110 mm
Katalog # WHAT1002-10240
Cone folded filter papers, Whatman™
Katalog # WHAT1002-10240
Cone folded filter papers, Whatman™
Cone folded filter papers, Whatman™
2, 110 mm
Price
1.860,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents
Cone folded filter paper saves valuable time when undertaking repetitive or multiple analyses.
  • Easily fits into conical funnels
  • Saves valuable time when undertaking repetitive or multiple analyses
  • Provides the same high quality and consistency that you expect from all Whatman filter papers
Whatman cellulose filter paper grades are available in ready-to-use cone folded formats.
Specifikationer
  • Kvalitet:
    2
  • Seq:
    02
  • Størrelse:
    110 mm
  • Tykkelse:
  • Cat. No.:
    WHAT1002-10240
  • Grundvægt:
  • Askeindhold: