Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human IgM ELISA Kit

Human IgM ELISA Kit

Human IgM ELISA kit, 10×96 tests
Katalog # ABCAAB214568-10X96
Leverandør:   Abcam
Human IgM ELISA Kit
Katalog # ABCAAB214568-10X96
Leverandør:  Abcam
CAS nummer:  
Human IgM ELISA Kit
Human IgM ELISA Kit

Human IgM ELISA kit, 10×96 tests

CAS nummer:  
Price
57.730,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Human IgM ELISA Kit is a single-wash 90-min Simplestep used to quantify Human IgM with a sensitivity of 50 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput. - Cited in over 5 citations.

Specifikationer

Specifikationer

  • Beskrivelse:
    Human IgM ELISA kit
  • Test pr. sæt:
    10×96 tests
  • Sensitivitet:
    50 pg/ml
  • Assay varighed:
    One step
  • Primary antibody reactivity:
    Human
  • Detektionsområde:
    390,63 - 25000 pg/ml
  • Prøvetype:
    Saliva, Heparin Plasma, Citrate plasma, Cell culture supernatant, Milk, Serum, EDTA Plasma
  • Time to Results:
    1 h 30 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ABCAAB214568-10X96
  • Format:
    Pre-coated
  • Target protein:
    IgM
  • Detektionsmetode:
    Colorimetric
  • Opbevaringstemperatur:
    +4 °C
  • Regulatory status:
    RUO

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.