Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Winter Jackets, PW2

Winter Jackets, PW2

marineblå/mørkeblå, XXL, Regular
Katalog # PRTWPW260NRRXXL
Leverandør:   Portwest
Winter Jackets, PW2
Katalog # PRTWPW260NRRXXL
Leverandør:  Portwest
Winter Jackets, PW2
Winter Jackets, PW2

marineblå/mørkeblå, XXL, Regular

Price
560,00 kr
Log ind for tilgængelighed
1 * 1 Par
Product Details & Documents

Stylish, functional, and durable, the PW2 winter jacket offers protection and warmth in extreme weather conditions. Key features include contrast coloured panels for added style, an adjustable detachable hood, and plenty of pockets for storage.

  • Quilt lined for thermal insulation
  • Non-magnetic, nickel and ferrous free
  • Internal chest pocket
  • Contrast colouring for added style
  • Print access for corporate branding
  • Radio loop for easy clipping of a radio
  • Hook and loop cuffs for a secure fit
  • EN 342 Protection against cold certified up to −40 °C

Waterproof jacket with taped seams preventing water penetration. Extremely water resistant eco-conscious fabric finish, the material beads water away from fabric surface.

 : EN 343 Class 3:1 X WP 5,000mm. EN 342 0.336M². K/W (B), 2, X. CE certified.

Documents

Specifikationer
  • Tøj størrelse:
    XXL
  • Beklædningsgenstand:
    Regular
  • Farve:
    marineblå/mørkeblå
  • Seq:
    17
  • Køn:
    For mænd
  • Cat. No.:
    PRTWPW260NRRXXL
  • Sleeve or leg length:
    Long
  • Risk area:
    Rain (waterproof)
  • Vægt:
    190 g/m²
  • Outfit applikationer:
    Outdoor
  • Lukkestørrelse:
    Med Velcro®
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.