BERLIN Stretch Trousers
Stretch, blå, L (56), Regular, 96 - 100 cm
Catalog # ALSW1035887-BE50
Supplier: ALSICO
Jump to:
Product Details & Documents
Functional men's trousers for the logistic sector that offer a professional look without compromising comfort. The fabric is powered by Alsiflex technology, which provides a unique combination of stretch, sustainability, and performance.
- Waistband with elastic in the side
- Front closure with jeans button and zipper
- 2 spacious side pockets
- Big open thigh pocket on right leg for scanner or tablet
- Left leg thigh pocket with pen pocket
- 2 back pockets
Industrial launderable.
Certifications: Oeko-Tex® standard 100.
Documents
International Body measurements - Alsico size chart
Additional Product Details
Specifications
- Tøj størrelse:L (56)
- Beklædningsgenstand:Regular
- Model:Stretch
- Farve:blå
- Seq:21
- Waist circumference:96 - 100 cm
- Køn:For mænd
- Cat. No.:ALSW1035887-BE50
- Sleeve or leg length:Long
- Vægt:265 g/m²
- Outfit applikationer:Industrial|Mechanical protection
- Lukkestørrelse:Med lynlås
- Stof:Alsiflex (79% polyester, 21% cotton)