PolyCHECK® Wipes
sort, 228,6×228,6 mm
Katalog # FOAMHT5291K-5
Leverandør: FOAMTEC
Gå til:
Product Details & Documents
PolyCHECK® Wipe, black polyester inspection wipe with sealed edge.
Specifikationer
- Farve:sort
- L×B:228,6×228,6 mm
- Seq:01