Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Deep Water Samplers

Deep Water Samplers

Deep water sampler, 1000 ml, 280 mm, Plast
Katalog # SAMP5079A-1000
Leverandør:   Sampling Systems
Deep Water Samplers
Katalog # SAMP5079A-1000
Leverandør:  Sampling Systems
Deep Water Samplers
Deep Water Samplers

Deep water sampler, 1000 ml, 280 mm, Plast

Price
7.360,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

The Deep water sampler can take samples of liquids from large depths.

  • Transparent plastic construction enables easy viewing
  • Ideal for sampling at large depths in rivers, lakes and large containers
  • Simple operation

The Deep water sampler is available in different sizes, it takes a sample at the point you start pulling the handle upwards. The simple operation means you lower to the required depth then pull it out.

Specifikationer
  • Kapacitet:
    1000 ml
  • Beskrivelse:
    Deep water sampler
  • Længde:
    280 mm
  • Materiale:
    Plast
  • Seq:
    01