Orders LinkContactUsLinkComponent
Hand Scrapers
Hand Scrapers
Hand scraper, white
Catalog # SAMP8336P-1
Supplier:   Sampling Systems
Hand Scrapers
Catalog # SAMP8336P-1
Supplier:  Sampling Systems
Hand Scrapers
Hand Scrapers
Hand scraper, white
Price
253,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These robust hand scrapers are perfect for scraping large areas.
  • Made from high quality PP
  • Robust and incorporates a large sturdy handle
  • Large 250 mm wide blade
  • Comes in three distinctive colours
The range of robust and sturdy hand scrapers from Sampling Systems are perfect for easily scraping large areas due to their large blade size and robust handle. The performance is further enhanced by the fact it is made out of PP giving it very good chemical resistance.
Specifications
  • Beskrivelse:
    Hand scraper, white
  • Seq:
    03