Spring til Overskrift
Spring til Hovedindhold
Spring til Sidefod
Udvidet søgning
Udvidet søgning
SDS
Certifikater
Producentdata
0
0,00 kr
Order Entry
Log ind
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Ordreindtastning
Ordrestatus
Din konto
Kurv
Denmark
Ordreindtastning
Ordrestatus
Din konto
Kurv
ContactUsLinkComponent
[EN]5 COMPONENT 1 * 100 mL
Katalog # CMPLRM056760L1
Leverandør:
CUSTOM MADE CHEMICALS LAB
Udvid billedet
[EN]5 COMPONENT 1 * 100 mL
Katalog # CMPLRM056760L1
Leverandør:
CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
Leverandør: VWR Chemicals
[EN]BUFFER A CX-1 PH 5.6 1 * 500 mL
Leverandør: Thermo Fisher Scientific
Indigo 95% (dye content), Sigma-Aldrich®
Leverandør: SIGMA ALDRICH MICROSCOPY